Acid red 52

Lieferant: Spectrum Chemical

A1586-5KGBL
SPCMA1586-5KGBLEA 5110 EUR
SPCMA1586-5KGBL
Acid red 52
Acid red 52

Acid Red 52 is a fluorescent rhodamine dye also known as Sulforhodamine B and Kiton Red, and used as a polar tracer.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: C₂₇H₃₀N₂O₇S₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00010180
CAS-Nummer: 3520-42-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR