Aluminium acetate basic
Lieferant: Spectrum Chemical
Aluminum Acetate Monobasic is a salt that is used as a topical astringent to treat skin irritations from poison ivy, substances such as soaps and cosmetics and insect bites. Aluminum acetate is particularly helpful for foot problems including athlete’s foot, swelling, and excessive sweating. It is also used to treat ear canal infections. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Formel:
C₄H₇AlO₅ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008688 CAS-Nummer: 142-03-0 EINECS: 205-518-2 |
Spezifikation Testergebnisse
Assay (%) | 28 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...