α-Acetyl-γ-butyrolactone
Lieferant: Spectrum Chemical
Synonyme:
alpha-Acetyl-gamma-hydroxybutyric acid Lactone, α-Acetyl-γ-hydroxybutyric acid Lactone, 3-Acetyldihydrofuran-2(3H)-one, 2-Acetyl-γ-butyrolactone, 2-Acetylbutyrolactone, alpha-Acetyl-gamma-butyrolactone, α-Acetylbutyrolactone, 3-Acetyl-4,5-dihydro-2(3H)-furanone
A2107-200LTBL
SPCMA2107-200LTBLEA
0
EUR
SPCMA2107-200LTBL
α-Acetyl-γ-butyrolactone
α-Acetyl-γ-butyrolacton
Formel:
C₆H₈O₃ Molecular Weight: 128.13 g/mol Dichte: 1,190 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005394 CAS-Nummer: 517-23-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...