4-Aminophenol ≥98%

Lieferant: Spectrum Chemical

Synonyme: 4-Hydroxyanilin

A2305-50KGBL
SPCMA2305-50KGBLEA 11880 EUR
SPCMA2305-50KGBL
4-Aminophenol ≥98%
4-Aminophenol

p-Aminophenol, also known as 4-Aminophenol, is a building block compound and can be used as a film developer.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: H₂NC₆H₄OH
Molecular Weight: 109,13 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007869
CAS-Nummer: 123-30-8
UN: 2512
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7af17b754d4ebaa0993b4544d4e6ab6c116724c&vid=6275667c2624ee686f61715acd420dc596e076fa&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay 98% min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR