Nɑ-Acetyl-L-glutamine
Lieferant: Spectrum Chemical
Synonyme:
N-Acetyl-L-glutamine, N-Ac-Gln-OH, Ac-L-Gln-OH, Ac-Gln-OH
A2393-100KGBL
SPCMA2393-100KGBLEA
0
EUR
SPCMA2393-100KGBL
Nɑ-Acetyl-L-glutamine
Nalpha-Acetyl-L-glutamine
N-Acetyl-L-glutamine is used as a skin conditioning agent.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formel:
C₇H₁₂N₂O₄ Molecular Weight: 188.18 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00038159 CAS-Nummer: 2490-97-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...