1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)-1,3-propanedione

Lieferant: Spectrum Chemical

Synonyme: Avobenzone, 1-(4-(tert-Butyl)phenyl)-3-(4-methoxyphenyl)propane-1,3-dione

A2682-25GM
SPCMA2682-25GMEA 385 EUR
SPCMA2682-25GM
1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)-1,3-propanedione
1-(4-tert-Butylphenyl)-3-(4-methoxyphenyl)-1,3-propanedione

Avobenzone is an oil soluble ingredient used in sunscreen products to absorb the full spectrum of UVA rays. Its ability to absorb ultraviolet light over a wider range of wavelengths than many organic sunscreen agents has led to its use in many commercial preparations marketed as 'broad spectrum' sunscreens. Avobenzone has an absorption maximum of 357 nm. It appears as an odorless, yellowish solid.

Formel: C₂₀H₂₂O₃
Molecular Weight: 310,39 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00210252
CAS-Nummer: 70356-09-1

Bestellen

Spezifikation Testergebnisse

Appearance Slightly Yellow-
Assay 97.0 % min.
Melting Point 81 - 86°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR