Allantoin ≥98,5% USP

Lieferant: Spectrum Chemical

Synonyme: 1-(2,5-Dioxoimidazolidin-4-yl)urea

A2721-25KGBL A2721-50KGBL A2721-500GM A2721-2.5KG
SPCMA2721-25KGBLEA 8090 EUR
SPCMA2721-25KGBL SPCMA2721-50KGBL SPCMA2721-500GM SPCMA2721-2.5KG
Allantoin ≥98,5% USP
Allantoin

Allantoin, USP is a moisturising ingredient.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: C₄H₆N₄O₃
Molecular Weight: 158.12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005260
CAS-Nummer: 97-59-6

Bestellen

Spezifikation Testergebnisse

Assay 98.5 - 101.0%
Acidity or Alkalinity To Pass Test
Loss On Drying max. 0.1 %
Organic Impurities max. 0.5 %
Residue On Ignition (Sulphated Ash) max. 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR