Potassium-L-aspartate dihydrate

Lieferant: Spectrum Chemical

Synonyme: L-Aspartic Acid Potassium Salt dihydrate, L-Asparaginic Acid Potassium Salt dihydrate

A3204-50KGBL
SPCMA3204-50KGBLEA 6390 EUR
SPCMA3204-50KGBL
Potassium-L-aspartate dihydrate
Potassium-L-aspartate dihydrate

L-Aspartic Acid Potassium Salt, Dihydrate, also known as Aminosuccinic acid, is an alpha-amino acid and considered to be pervasive in biosynthesis; it is the precursor to many amino acids in microorganisms and plants.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: C₄H₆NO₄·K·2H₂O
Molecular Weight: 207.223 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00021083
CAS-Nummer: 1115-63-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR