Adipohydrazide ≥99%

Lieferant: Spectrum Chemical

Synonyme: Adipyl hydrazide

A3802-20KGBL
SPCMA3802-20KGBLEA 4820 EUR
SPCMA3802-20KGBL
Adipohydrazide ≥99%
Adipohydrazid

Adipic Acid Dihydrazide, also known as ADH or Adipohydrazide, can be used as a hardener for epoxy resins and for cross-linking water-based emulsions. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formel: C₆H₁₄N₄O₂
Molecular Weight: 174.2 g/mol
Siedepunkt: 446 °C (1013 hPa)
Schmelzpunkt: 181…182 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007614
CAS-Nummer: 1071-93-8
EINECS: 213-999-5

Bestellen

Spezifikation Testergebnisse

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR