Aminoguanidine hydrochloride

Lieferant: Spectrum Chemical

Synonyme: Aminoguanidine HCl, Aminoguanidine Hydrochloride, Guanylhydrazine hydrochloride

A3814-40KGBL
SPCMA3814-40KGBLEA 0 EUR
SPCMA3814-40KGBL
Aminoguanidine hydrochloride
Aminoguanidin Hydrochlorid

Aminoguanidine Hydrochloride, also known as Pimagedine or Guanyl hydrazine, is a nitric oxide and diamine oxidase synthase inhibitor which can be used for energetic material applications.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: CH₇ClN₄
Molecular Weight: 110,55 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00039074
CAS-Nummer: 1937-19-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR