Adenosine-5'-triphosphate disodium salt (ATP disodium salt) ≥99%
Lieferant: Spectrum Chemical
Synonyme:
((2R,3S,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl tetrahydrogen triphosphate, disodium salt, Adenosine 5'-triphosphoric acid disodium, Adenosine 5′-triphosphate disodium salt, ATP 2Na, ATP disodium salt, Adenosine 5ʹ-Triphosphate, Disodium Salt
Adenosine-5'-triphosphate Disodium Salt is a nucleoside triphosphate that cells use as a coenzyme. It is used by structural proteins and enzymes in several types of cellular processes that include motility and biosynthetic reactions.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formel:
C₁₀H₁₄N₅Na₂O₁₃P₃ Molecular Weight: 551,14 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00080339 CAS-Nummer: 987-65-5 EINECS: 213-579-1 |
Spezifikation Testergebnisse
Assay (%) | 99 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...