Agar, powder NF

Lieferant: Spectrum Chemical

Synonyme: Agar Agar

AG110-2.5KG AG110-25KGBL
SPCMAG110-2.5KGEA 1220 EUR
SPCMAG110-2.5KG SPCMAG110-25KGBL
Agar, powder NF
Agar

Form: Powder

Schmelzpunkt: 90
Lagertemperatur: Raumtemperatur
MDL: MFCD00081288
CAS-Nummer: 9002-18-0
EINECS: 232-658-1
Merck Index: 13,00184
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=36877cbcfaca6a0ace648ee20ae51b3467f2748c&vid=4295a0dcbc54bbbd374a5fb934297574c6848d32&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR