AMP (2-Amino-2-methylpropanol)

Lieferant: Spectrum Chemical
Achtung

AM156-200LTBL
SPCMAM156-200LTBLEA 9710 EUR
SPCMAM156-200LTBL
AMP (2-Amino-2-methylpropanol)
AMP (2-Amino-2-methylpropanol)

2-Amino-2-methyl-1-propanol, or aminomethyl propanol is a colourless, viscous liquid that functions as a pH adjuster. It is also used as an intermediate in drug synthetic schemes.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: (CH₃)₂C(NH₂)CH₂OH
Siedepunkt: 165 °C (1013 hPa)
Schmelzpunkt: 18…26 °C
Dichte: 0,934
Flammpunkt: 67 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008051
CAS-Nummer: 124-68-5
EINECS: 204-709-8
Merck Index: 13,00448

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR