Barium peroxide, reagent

Lieferant: Spectrum Chemical

Synonyme: Bariumsuperoxid

B1053-12KGBL
SPCMB1053-12KGBLEA 3850 EUR
SPCMB1053-12KGBL
Barium peroxide, reagent
Bariumperoxid

Barium Peroxide, Powder, Reagent is an extremely powerful oxidising agent, and therefore commonly used in explosives and pyrotechnics. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formel: BaO₂
Molecular Weight: 169.33 g/mol
Schmelzpunkt: 450
Lagertemperatur: Raumtemperatur
MDL: MFCD00003454
CAS-Nummer: 1304-29-6
UN: 1449
ADR: 5.161 II
Merck Index: 12,01018

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR