Benzyltriethylammonium chloride
Lieferant: Spectrum Chemical
|
Achtung
|
Synonyme:
Benzyl triethylammonium chloride
B1083-45KGBL
SPCMB1083-45KGBLEA
8930
EUR
SPCMB1083-45KGBL
Benzyltriethylammonium chloride
Benzyltriethylammoniumchlorid
Benzyltriethylammonium Chloride, also known as Triton B, is a quaternary ammonium salt that functions as an organic base and is a popular phase-transfer catalyst.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formel:
C₁₃H₂₂ClN Molecular Weight: 227,78 g/mol Schmelzpunkt: 190…192 °C Flammpunkt: >100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011824 CAS-Nummer: 56-37-1 EINECS: 200-270-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...