Bis(2-methoxyethyl)ether ≥99%, Reagenzqualität

Lieferant: Spectrum Chemical
Gefahr

Synonyme: Dimethyldiglycol, 2-Methoxyethylether, Diethylenglycoldimethylether, Dimethyldigol, Diglyme

B1099-200LTBL
SPCMB1099-200LTBLEA 0 EUR
SPCMB1099-200LTBL
Bis(2-methoxyethyl)ether ≥99%, Reagenzqualität
Bis(2-methoxyethyl)ether

Bis(2-methoxyethyl) Ether, Reagent is more commonly referred to as Diglyme and is an organic compound that is a clear and colorless liquid with a faint ether like smell. It is solvent that has a high boiling point and that is miscible with water, alcohol as well as hydrocarbon solvents. Its main use is as a solvent for organic reactions as well as being able to chelate small cations. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Formel: C₆H₁₄O₃
Molecular Weight: 134,18 g/mol
Siedepunkt: 162 °C (1013 hPa)
Schmelzpunkt: –64 °C
Dichte: 0,937
Flammpunkt: 53 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008503
CAS-Nummer: 111-96-6
EINECS: 203-924-4
UN: 3271
ADR: 3,III
Merck Index: 13,03187

Bestellen

Spezifikation Testergebnisse

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR