1-Butanol NF

Lieferant: Spectrum Chemical
Gefahr

Synonyme: 1-Butylalkohol, 1-Hydroxybutan, n-Butanol, Propylcarbinol

B1197-1LT B1197-20LT B1197-200LTBL
SPCMB1197-1LTEA 246 EUR
SPCMB1197-1LT SPCMB1197-20LT SPCMB1197-200LTBL
1-Butanol NF
1-Butanol

Butyl Alcohol, NF is primarily a solvent.


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).

Formel: H₃C(CH₂)₂CH₂OH
Siedepunkt: 117,6 °C (1013 hPa)
Schmelzpunkt: –89 °C
Dichte: 0,811
Flammpunkt: 30 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002964
CAS-Nummer: 71-36-3
EINECS: 200-751-6
UN: 1120
ADR: 3,III
REACH: 01-2119484630-38
Merck Index: 13,01539
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9395eeb6150a74222da52460f53cedeccd93442&vid=fd8352a556914c5be7213c576355a88e130d1a7b&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR