Benzyl nicotinate
Lieferant: Spectrum Chemical
|
Achtung
|
Synonyme:
Benzyl-3-pyridincarboxylat, Pyridin-3-carbonsäurebenzylester, 3-Picolinsäurebenzylester, 3-Pyridincarbonsäurebenzylester, Nicotinsäurebenzylester
B1212-1KGBL
SPCMB1212-1KGBLEA
4530
EUR
SPCMB1212-1KGBL
Benzyl nicotinate
Benzylnicotinat
Benzyl Nicotinate is a reactant used in the synthesis of indolizine derivatives.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formel:
C₁₃H₁₁NO₂ Siedepunkt: 179…180 °C (8 hPa) Schmelzpunkt: 24 °C Dichte: 11,165 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00023584 CAS-Nummer: 94-44-0 EINECS: 202-332-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...