Oxybenzone ≥97% USP

Lieferant: Spectrum Chemical

Synonyme: Eusolex® 4360, 2-Hydroxy-4-methoxybenzophenon

B1261-25GM
SPCMB1261-25GMEA 148 EUR
SPCMB1261-25GM
Oxybenzone ≥97% USP
Oxybenzon

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: C₁₄H₁₂O₃
Molecular Weight: 228,25 g/mol
Siedepunkt: 370 °C (1013 hPa)
Schmelzpunkt: 65 °C
Dichte: 1,3397 g/cm³ (25 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008387
CAS-Nummer: 131-57-7
EINECS: 205-031-5
Merck Index: 12,07088

Bestellen

Spezifikation Testergebnisse

Assay 97% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR