Benzyl acetate ≥98%
Lieferant: Spectrum Chemical
Benzyl Acetate, an organic compound, is the ester formed by condensation of acetic acid and benzyl alcohol; it is the main constituent of the essential oils from jasmine flowers.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formel:
C₉H₁₀O₂ Siedepunkt: 206 °C (1013 hPa) Schmelzpunkt: –51 °C Dichte: 1,04 Flammpunkt: 95 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008712 CAS-Nummer: 140-11-4 EINECS: 205-399-7 Merck Index: 13,01125 |
Spezifikation Testergebnisse
Assay | 98% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...