Benzidin
Lieferant: Spectrum Chemical
|
Gefahr
|
Synonyme:
4,4'-Diaminobiphenyl, 1,1’-Biphenyl-4,4’-diamin, 4,4′-Bisanilin, Biphenyl-4,4'-ylendiamin, Paradiaminobiphenyl, 4-(4-Aminophenyl)anilin
BB2010-1GM
SPCMBB2010-1GMEA
0
EUR
SPCMBB2010-1GM
Benzidin
Benzidin
Benzidine, also known as 4,4'-diaminobiphenyl, is a component of a test for cyanide. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formel:
C₁₂H₁₂N₂ Molecular Weight: 184,24 g/mol Siedepunkt: 401 °C Schmelzpunkt: 127 °C |
MDL:
MFCD00039765 CAS-Nummer: 92-87-5 UN: 1885 ADR: 6.1,II |
Spezifikation Testergebnisse
ASSAY | APPROX. 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...