Aktivkohle Coconut
Lieferant: Spectrum Chemical
Synonyme:
Carbon activated, Activated carbon, Activated charcoal
C1221-500GM
SPCMC1221-500GMEA
359
EUR
SPCMC1221-500GM
Aktivkohle Coconut
Aktivkohle
Formel:
C Molecular Weight: 12.01 g/mol Schmelzpunkt: 3550 |
MDL:
MFCD00133992 CAS-Nummer: 7440-44-0 Merck Index: 13,01818 |
Spezifikation Testergebnisse
MESH SIZE | 8 - 30 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...