Copper(II) gluconate

Lieferant: Spectrum Chemical

C1334-2.5KG C1334-25KGBL
SPCMC1334-2.5KGEA 1940 EUR
SPCMC1334-2.5KG SPCMC1334-25KGBL
Copper(II) gluconate
Copper(II) gluconate
Formel: C₁₂H₂₂CuO₁₄
MDL: MFCD00075297
CAS-Nummer: 527-09-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR