Chlorophyllin, coppered trisodium salt, Pulver

Lieferant: Spectrum Chemical

C1443-12KGBL
SPCMC1443-12KGBLEA 0 EUR
SPCMC1443-12KGBL
Chlorophyllin, coppered trisodium salt, Pulver
Chlorophyllin, coppered trisodium salt

Chlorophyllin, Powder, also known as Natural Green 3, E141, and is a water soluble salt that is a semi-synthetic derivative of chlorophyll.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: C₃₄H₃₁CuN₄Na₃O₆
Molecular Weight: 724,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012149
CAS-Nummer: 11006-34-1

Bestellen

Spezifikation Testergebnisse

Content of Chelated Copper 4.0% min.
Content of Total Copper 4.25 % min.
pH (1% in Water) 9.0 - 10.9
Ash max. 36 %
Arsenic max. 3 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR