α-Cyclodextrin 98
Lieferant: Spectrum Chemical
C1620-5KGBL
SPCMC1620-5KGBLEA
26580
EUR
SPCMC1620-5KGBL
α-Cyclodextrin 98
α-Cyclodextrin
alpha-Cyclodextrin, a polysaccharide, can be used as a dietary fiber.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formel:
C₃₆H₆₀O₃₀ Molecular Weight: 972.84 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00078207 CAS-Nummer: 10016-20-3 EINECS: 233-007-4 |
Spezifikation Testergebnisse
Assay (%) | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...