Calcium thioglycolate trihydrate

Lieferant: Spectrum Chemical

Synonyme: Calciumbis(mercaptoacetat) Trihydrat

C1701-50KGBL
SPCMC1701-50KGBLEA 0 EUR
SPCMC1701-50KGBL
Calcium thioglycolate trihydrate
Calciumthioglycolat Trihydrat

Calcium Thioglycolate, Trihydrate, also known as Acetyl mercaptan or Thioglycolic acid, is used as a chemical depilatory.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: C₄H₆CaO₄S₂·3H₂O
Molecular Weight: 276,35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00150707
CAS-Nummer: 65208-41-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR