Calciumchlorid, wasserfrei ≥93,0%, Plätzchen FCC

Lieferant: Spectrum Chemical

Synonyme: Chlorcalcium

C1977-500GM
SPCMC1977-500GMEA 128 EUR
SPCMC1977-500GM
Calciumchlorid, wasserfrei ≥93,0%, Plätzchen FCC
Calciumchlorid

Calcium Chloride, Anhydrous, Pellets, FCC is used as a sequestrant and firming agent in food products. It is considered as generally recognized as safe (GRAS) by the U.S. Food and Drug Administration. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark.

Formel: CaCl₂
Molecular Weight: 110.98 g/mol
Siedepunkt: 1600
Schmelzpunkt: 772
Lagertemperatur: Raumtemperatur
MDL: MFCD00010903
CAS-Nummer: 10043-52-4
Merck Index: 14,01659

Bestellen

Spezifikation Testergebnisse

Assay 93.0 - 100.5 %
Acid-Insoluble Substances max. 0.02 %
Arsenic (As) max. 3 mg/kg
Fluoride max. 0.004 %
Lead (Pb) max. 5 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR