Cyclohexylamine hydrochloride

Lieferant: Spectrum Chemical

Synonyme: Cyclohexylamine Hydrochloride

C2127-25KGBL
SPCMC2127-25KGBLEA 0 EUR
SPCMC2127-25KGBL
Cyclohexylamine hydrochloride
Cyclohexylamine hydrochloride

Cyclohexylamine Hydrochloride is an organic compound that is an aliphatic amine. The compound is a high-melting solid. Cyclohexylamine hydrochloride is used as an intermediate in the synthesis of other organic compounds.

Formel: C₆H₁₃N
Molecular Weight: 135,64 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00050540
CAS-Nummer: 4998-76-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR