Cumene hydroperoxide

Lieferant: Spectrum Chemical

Synonyme: Cumyl hydroperoxide

C2313-200LTBL
SPCMC2313-200LTBLEA 0 EUR
SPCMC2313-200LTBL
Cumene hydroperoxide
Cumolhydroperoxid

Cumene Hydroperoxide, also known as Cumyl Hydroperoxide, is used as an oxidising agent.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: C₉H₁₂O₂
Molecular Weight: 152.20 g/mol
Dichte: 1,038
Lagertemperatur: Kühlschrank
MDL: MFCD00002129
CAS-Nummer: 80-15-9
UN: 3109
ADR: 5.2 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR