Calciumsulfat Dihydrat ≥98,0% FCC

Lieferant: Spectrum Chemical

Synonyme: Pflaster Paris

CA173-12KG CA173-2.5KG CA173-500GM
SPCMCA173-12KGEA 1060 EUR
SPCMCA173-12KG SPCMCA173-2.5KG SPCMCA173-500GM
Calciumsulfat Dihydrat ≥98,0% FCC
Calciumsulfat Dihydrat

Calcium Sulfate, Dihydrate, FCC is widely used as pH regulator, hardness regulator, yeast agent, nutritional suppliment, texture and flavor enhancer in food production. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formel: CaSO₄·2H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00149625
CAS-Nummer: 10101-41-4

Bestellen

Spezifikation Testergebnisse

Assay (Dried Basis) 98.0 % min.
Loss On Drying 19.0 - 23.0 %
Fluoride max. 0.003 %
Lead (Pb) max. 2 mg/kg
Selenium (Se) max. 0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR