trans-Cinnamaldehyde ≥98%, Reagenzqualität

Lieferant: Spectrum Chemical

Synonyme: trans-3-Phenyl-2-propenal

CI110-200LTBL
SPCMCI110-200LTBLEA 14700 EUR
SPCMCI110-200LTBL
trans-Cinnamaldehyde ≥98%, Reagenzqualität
trans-Zimtaldehyd

trans-Cinnamaldehyde, Reagent, also known as Cinnamic aldehyde and trans-cinnamaldehyde, is used as a corrosion inhibitor for ferrous alloys and steel in corrosive fluids. The Reagent grade denotes that this chemical is the highest quality commercially available and that the American Chemical Society has not officially set any specifications for this material. Spectrum Chemical manufactured Reagent grade products meet the toughest regulatory standards for quality and purity.

Formel: C₉H₈O
Molecular Weight: 132,16 g/mol
Siedepunkt: 252 °C (1013 hPa)
Schmelzpunkt: –8 °C
Dichte: 1,05 g/cm³ (20 °C)
Flammpunkt: 138 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00007000
CAS-Nummer: 14371-10-9
Merck Index: 12,02319

Bestellen

Spezifikation Testergebnisse

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR