Acid Red 18 (Food Red No. 102)

Lieferant: Spectrum Chemical

Synonyme: Cochineal Red A, New Coccine, Scarlet 3R, Sodium 7-hydroxy-8-((4-sulphonatonaphthalen-1-yl)diazenyl)naphthalene-1,3-disulphonate, Ponceau 4R, Cochenille red

CO102-50KGBL
SPCMCO102-50KGBLEA 0 EUR
SPCMCO102-50KGBL
Acid Red 18 (Food Red No. 102)
Acid Red 18 (Food Red No. 102)

Cochineal Red A, also known as ponceau 4R, is typically synthesized from aromatic hydrocarbons and used as a synthetic food coloring. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: C₂₀H₁₄N₂O₁₀S₃
Molecular Weight: 604,46 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004084
CAS-Nummer: 2611-82-7

Bestellen

Spezifikation Testergebnisse

Absorption Max 506 (335) NM

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR