Coconut oil, raffiniert

Lieferant: Spectrum Chemical

CO110-12KG CO110-2.5KG CO110-200LTBL
SPCMCO110-12KGEA 733 EUR
SPCMCO110-12KG SPCMCO110-2.5KG SPCMCO110-200LTBL
Coconut oil, raffiniert
Coconut oil

Coconut Oil, Refined is taken from the meat of coconuts that come from the coconut palm. It is considered an edible oil and has several purposes in food, industry, and medicine. It contains high saturated fat.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Lagertemperatur: Raumtemperatur
MDL: MFCD00130817
CAS-Nummer: 8001-31-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR