Baumwollsamenöl NF
Lieferant: Spectrum Chemical
CO145-200LTBL
CO145-20LT
CO145-4LTGL
CO145-500MLGL
SPCMCO145-200LTBLEA
3600
EUR
SPCMCO145-200LTBL
SPCMCO145-20LT
SPCMCO145-4LTGL
SPCMCO145-500MLGL
Baumwollsamenöl NF
Cottonseed oil
Dichte:
0,91 g/cm³ (20 °C) |
MDL:
MFCD00130872 CAS-Nummer: 8001-29-4 EINECS: 232-280-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...