Diethanolamine ≥98,5% ACS

Lieferant: Spectrum Chemical
Gefahr

Synonyme: 2,2'-Iminodiethanol

D1033-500MLGL
SPCMD1033-500MLGLEA 196 EUR
SPCMD1033-500MLGL
Diethanolamine ≥98,5% ACS
Diethanolamin

Diethanolamine, Reagent, ACS is also referred to as DEA, is an organic compound that in its pure form is a white solid but it is most often encountered as a clear liquid that has a viscous texture. It is a secondary amine and diol and is water soluble. It is used as a corrosion inhibitor and surfactant that is used in the oil refining industry to help remove hydrogen sulfide from sour gas. As an ACS grade reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness

Formel: C₄H₁₁NO₂
Molecular Weight: 105,14 g/mol
Siedepunkt: 268,8 °C (1013 hPa)
Schmelzpunkt: 28 °C
Dichte: 1,088
Flammpunkt: 176 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002843
CAS-Nummer: 111-42-2
EINECS: 203-868-0
REACH: 01-2119488930-28
Merck Index: 13,03134

Bestellen

Spezifikation Testergebnisse

Assay 98.5% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR