1,3-Dihydroxyacetone dimer ≥98%

Lieferant: Spectrum Chemical

Synonyme: 2,5-Dihydroxy-1,4-dioxane-2,5-dimethanol

D1038-1KG D1038-25KGBL
SPCMD1038-1KGEA 912 EUR
SPCMD1038-1KG SPCMD1038-25KGBL
1,3-Dihydroxyacetone dimer ≥98%
1,3-Dihydroxyaceton dimer

Dihydroxyacetone, Dimer, also known as glycerone, is used as an ingredient in sunless tanning products and can be derived from the fermentation of glycerin.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: C₆H₁₂O₆
Molecular Weight: 180.16 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00051019
CAS-Nummer: 62147-49-3

Bestellen

Spezifikation Testergebnisse

Assay 98% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR