1,3-Dihydroxyacetone dimer ≥98%
Lieferant: Spectrum Chemical
|
|
Synonyme:
2,5-Dihydroxy-1,4-dioxane-2,5-dimethanol
D1038-1KG
D1038-25KGBL
SPCMD1038-1KGEA
912
EUR
SPCMD1038-1KG
SPCMD1038-25KGBL
1,3-Dihydroxyacetone dimer ≥98%
1,3-Dihydroxyaceton dimer
Dihydroxyacetone, Dimer, also known as glycerone, is used as an ingredient in sunless tanning products and can be derived from the fermentation of glycerin.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formel:
C₆H₁₂O₆ Molecular Weight: 180.16 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00051019 CAS-Nummer: 62147-49-3 |
Spezifikation Testergebnisse
Assay | 98% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...