Bis(2-ethylhexyl)adipate
Lieferant: Spectrum Chemical
|
Achtung
|
Synonyme:
Di-(2-ethylhexyl) adipate, Bis(2-ethylhexyl) hexanedioate, Dioctyl Adipate, Diisooctyl adipate, Bis(2-ethylhexyl) adipate
D1246-200LTBL
SPCMD1246-200LTBLEA
4000
EUR
SPCMD1246-200LTBL
Bis(2-ethylhexyl)adipate
Bis(2-ethylhexyl)adipat
Dioctyl Adipate is a diester which is used as a plasticizer.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formel:
C₂₂H₄₂O₄ Molecular Weight: 370,57 g/mol Dichte: 0,927 Lagertemperatur: Kühlschrank |
MDL:
MFCD00009496 CAS-Nummer: 103-23-1 EINECS: 203-090-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...