Diisopropanolamin

Lieferant: Spectrum Chemical
Achtung

Synonyme: 1,1'-Iminodipropan-2-ol, Bis(2-hydroxypropyl)amin

D1324-40LBBL
SPCMD1324-40LBBLEA 2060 EUR
SPCMD1324-40LBBL
Diisopropanolamin
Diisopropanolamin

Diisopropanolamine, also known as 1-amino-2-propanol or isopropanolamine, is an amino alcohol.


Used to achieve buffering, basicity, and alkalinity objectives.

Formel: C₆H₁₅NO₂
Molecular Weight: 133,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004531
CAS-Nummer: 110-97-4
EINECS: 203-820-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR