N-Ethyldiisopropylamine ≥99%

Lieferant: Spectrum Chemical

Synonyme: N,N-Diisopropylethylamin

D2163-200LTBL
SPCMD2163-200LTBLEA 28390 EUR
SPCMD2163-200LTBL
N-Ethyldiisopropylamine ≥99%
N-Ethyldiisopropylamin

N,N-Diisopropylethylamine is a colorless liquid and is also known as Hunig's base. N,N-DIISOPROPYLETHYLAMINE is a good base but a poor nucleophile. This characteristics make this material an extremely versatile synthetic organic reagent


GHS hazard : Flame;Corrosion;Skull and crossbones;Exclamation mark
UN2733

Formel: C₈H₁₉N
Molecular Weight: 129,25 g/mol
Dichte: 0,742
Lagertemperatur: Raumtemperatur
MDL: MFCD00008868
CAS-Nummer: 7087-68-5
UN: 2733
ADR: 38,II

Bestellen

Spezifikation Testergebnisse

Assay 0.99

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR