Dimethyl carbonate ≥99%

Lieferant: Spectrum Chemical
Gefahr

Synonyme: Kohlensäuredimethylester, Methylcarbonat

D2285-200LTBL D2285-20KG
SPCMD2285-200LTBLEA 4800 EUR
SPCMD2285-200LTBL SPCMD2285-20KG
Dimethyl carbonate ≥99%
Dimethylcarbonat

Dimethyl Carbonate is an organic compound that is a colorless and flammable liquid that is classified as a carbonate ester. It is considered to be a green methylating reagent and classified as a solvent that is exempt from classification as a volatile organic compound. It has also been considered as an alternative fuel oxygenate additive.

Formel: C₃H₆O₃
Siedepunkt: 90 °C (1013 hPa)
Schmelzpunkt: 0,5…4,7 °C
Dichte: 1,069
Flammpunkt: 18 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008420
CAS-Nummer: 616-38-6
EINECS: 210-478-4
UN: 1161
ADR: 3,II
Merck Index: 12,06117

Bestellen

Spezifikation Testergebnisse

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR