Capric acid ≥99%

Lieferant: Spectrum Chemical
Achtung

Synonyme: Decansäure

D2659-200LTBL
SPCMD2659-200LTBLEA 0 EUR
SPCMD2659-200LTBL
Capric acid ≥99%
Caprinsäure

Decanoic Acid is also known as capric acid. It is a saturated fatty acid from coconut oil and palm kernel oil. It can also be found in the milk of several types of mammals.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: C₁₀H₂₀O₂
Molecular Weight: 172,27 g/mol
Siedepunkt: 268 °C (1013 hPa)
Schmelzpunkt: 29…32 °C
Dichte: 0,893
Flammpunkt: 147 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004441
CAS-Nummer: 334-48-5
EINECS: 206-376-4
Merck Index: 13,01764

Bestellen

Spezifikation Testergebnisse

Assay 99% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR