Diazolidinyl urea ≥95%
Lieferant: Spectrum Chemical
|
|
Synonyme:
1,3-Bis(hydroxymethyl)-5-[1,3-bis(hydroxymethyl)ureido]hydantoin, 1-(1,3-Bis(hydroxymethyl)-2,5-dioxoimidazolidin-4-yl)-1,3-bis(hydroxymethyl)urea
Diazolidinyl Urea is an antimicrobial preservative most often used in cosmetics, skin care products, shampoos and conditioners, as well as a wide range of products including bubble baths, baby wipes and household detergents.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formel:
C₈H₁₄N₄O₇ Molecular Weight: 278,22 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD03547942 CAS-Nummer: 78491-02-8 |
Spezifikation Testergebnisse
Assay | Approx. 95% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...