Dodecyltrimethylammonium bromide

Lieferant: Spectrum Chemical

Synonyme: Dodecyl trimethyl ammonium bromide, (1-Dodecyl)trimethylammonium bromide

D3009-10KGBL
SPCMD3009-10KGBLEA 9620 EUR
SPCMD3009-10KGBL
Dodecyltrimethylammonium bromide
Dodecyltrimethylammonium bromide

Dodecyltrimethylammonium Bromide is a quaternary ammonium salt useful as paint strippers, foaming stabilizers, and bactericidal lotions.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: C₁₅H₃₄BrN
Schmelzpunkt: 264…266 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011767
CAS-Nummer: 1119-94-4
EINECS: 214-290-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR