Denatoniumbenzoat, anhydrous 99,5, granular NF

Lieferant: Spectrum Chemical

Synonyme: N-Benzyl-2-((2,6-dimethylphenyl)amino)-N,N-diethyl-2-oxoethanaminium benzoate

DE101-25GM DE101-5GM DE101-1KGBL
SPCMDE101-25GMEA 1380 EUR
SPCMDE101-25GM SPCMDE101-5GM SPCMDE101-1KGBL
Denatoniumbenzoat, anhydrous 99,5, granular NF
Denatoniumbenzoat

Denatonium Benzoate, Anhydrous, Granular, NF is used as a denaturant and masking agent.


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).

Formel: C₂₈H₃₄N₂O₃
Molecular Weight: 446.59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00031578
CAS-Nummer: 3734-33-6
EINECS: 223-095-2

Bestellen

Spezifikation Testergebnisse

Assay 99.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR