Dextrin, powder_white

Lieferant: Spectrum Chemical

Synonyme: Dextrin

DE137-2.5KG DE137-500GM
SPCMDE137-2.5KGEA 275 EUR
SPCMDE137-2.5KG SPCMDE137-500GM
Dextrin, powder_white
Dextrin

Dextrin, White Powder is used as water-soluble glue in envelope adhesives, crispness enhancers in food processing and as a thickening agent in pharmaceuticals.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: (C₆H₁₂O₆)n
Molecular Weight: 180.16 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00081554
CAS-Nummer: 9004-53-9
EINECS: 232-675-4
Merck Index: 13,02971

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR