EDTA Dinatriumsalz Dihydrat ≥99,0% FCC

Lieferant: Spectrum Chemical

Synonyme: Kestranal® 2S 2 H2O, Ethylendinitrilotetratetraessigsäure Dinatriumsalz Dihydrat, Dinatriumdihydrogenethylendiamintetraacetat Dihydrat, Titriplex III, Edetinsäure Dinatriumsalz Dihydrat, Ethylendiamin-N,N,N',N'-tetraessigsäure Dinatriumsalz Dihydrat, EDTA-Na2 Dihydrat, Kestranal® 2S Dihydrat, Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat

E1001-125GM E1001-500GM
SPCME1001-125GMEA 147 EUR
SPCME1001-125GM SPCME1001-500GM
EDTA Dinatriumsalz Dihydrat ≥99,0% FCC
EDTA Dinatriumsalz Dihydrat

Disodium EDTA, Dihydrate, FCC is used as a preservative to inhibit discoloration. Spectrum Chemical manufactured FCC products, including all FCC Disodium products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: C₁₀H₁₄N₂Na₂O₈·2H₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00150037
CAS-Nummer: 6381-92-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (C₁₀H₁₄O₈N₂Na₂2H₂O) 99.0 - 101.0 %
pH of a 1 in 100 Solution 4.3 - 4.7
Calcium (Ca) To Pass Test
Lead (Pb) max. 10 mg/kg
Nitrilotriacetic Acid[N[(HOCOCH₂)₃N] To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR