Ethyloleat NF

Lieferant: Spectrum Chemical

E1172-5KGBL E1172-100GM E1172-500GM
SPCME1172-5KGBLEA 0 EUR
SPCME1172-5KGBL SPCME1172-100GM SPCME1172-500GM
Ethyloleat NF
Ethyloleat

Ethyl Oleate, NF is used as a solubilizing agent in pharmaceutical preparations. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formel: C₂₀H₃₈O₂
Siedepunkt: 190…210 °C
Flammpunkt: 160 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009579
CAS-Nummer: 111-62-6

Bestellen

Spezifikation Testergebnisse

Assay (As Ethyl (Z)-9-Octadecenoate) To Pass Test
Presence of Ester To Pass Test
Iodine Value 75 - 85
Peroxide Value max. 10.0
Water Determination max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR