Ammonium iron (III) citrate, braun FCC

Lieferant: Spectrum Chemical

Synonyme: Ammonium ferric citrate, Ferric Ammonium Citrate, Ammonium iron (III) citrate

F1000-2.5KG F1000-500GM
SPCMF1000-2.5KGEA 698 EUR
SPCMF1000-2.5KG SPCMF1000-500GM
Ammonium iron (III) citrate, braun FCC
Ammoniumeisen(III)citrat

Ferric Ammonium Citrate, Brown, Powder, FCC is a brownish powder which is easily soluble in water. It is a food additive used as an acidity regulator and anti-caking agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: C₆H₅+4y)Fe(x)N(y)O₇
Siedepunkt: 197 °C (1013 hPa)
Dichte: 1,8 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00013099
CAS-Nummer: 1185-57-5
EINECS: 214-686-6

Bestellen

Spezifikation Testergebnisse

Assay 16.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR