Ammoniumeisen(III)citrat, grün, Pulver FCC

Lieferant: Spectrum Chemical

F1001-500GM F1001-2.5KG
SPCMF1001-500GMEA 198 EUR
SPCMF1001-500GM SPCMF1001-2.5KG
Ammoniumeisen(III)citrat, grün, Pulver FCC
Ammoniumeisen(III)citrat

Ferric Ammonium Citrate, Green, Powder, FCC is also known as Ferric ammonium citrate or Ammonium Iron(III) Citrate and is used as an acidity regulator. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: C₆H₅+4y)Fe(x)N(y)O₇
Lagertemperatur: Raumtemperatur
MDL: MFCD00013099
CAS-Nummer: 1185-57-5

Bestellen

Spezifikation Testergebnisse

Assay (Fe) 14.5 - 16.0 %
Ferric Citrate To Pass Test
Oxalate (C₂O₄) To Pass Test
Sulfate max. 0.3 %
Mercury max. 1 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR